accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121882-75-5

IUPAC Name: dichlorotris[4-(trifluoromethyl)phenyl]bismuthane
Synonyms: dichlorotris[4-(trifluoromethyl)phenyl]bismuthane
Molecular Weight (g/mol): 715.19
Molecular Formula: C21H12BiCl2F9
InChi Key: VOZWPVZQISDWJM-UHFFFAOYSA-L
SMILES: FC(F)(F)C1=CC=C(C=C1)[Bi](Cl)(Cl)(C1=CC=C(C=C1)C(F)(F)F)C1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results