accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121916-94-7

IUPAC Name: 41-amino-5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontan-31,32,33,34,35,36,37,38,39,40,42-undecol
Synonyms: 41-amino-5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontan-31,32,33,34,35,36,37,38,39,40,42-undecol
Molecular Weight (g/mol): 971.86
Molecular Formula: C36H61NO29
InChi Key: TXBHONGFOHSVMN-UHFFFAOYNA-N
SMILES: NC1C(O)C2OC(CO)C1OC1OC(CO)C(OC3OC(CO)C(OC4OC(CO)C(OC5OC(CO)C(OC6OC(CO)C(O2)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O)C(O)C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results