accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121933-59-3

IUPAC Name: {4-methyl-4H-thieno[3,2-b]pyrrol-5-yl}methanol
Synonyms: {4-methylthieno[3,2-b]pyrrol-5-yl}methanol
Molecular Weight (g/mol): 167.23
Molecular Formula: C8H9NOS
InChi Key: ZRWPUHUHJRQLCD-UHFFFAOYSA-N
SMILES: CN1C(CO)=CC2=C1C=CS2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results