accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOOFF

CAS RN 1219454-89-3

IUPAC Name: 2-difluoromethanesulfonylpyridine
Synonyms: 2-difluoromethanesulfonylpyridine
Molecular Weight (g/mol): 193.17
Molecular Formula: C6H5F2NO2S
InChi Key: YRQNSTAWTLXCEZ-UHFFFAOYSA-N
SMILES: FC(F)S(=O)(=O)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)