accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3H3C

CAS RN 122-19-0

IUPAC Name: benzyldimethyloctadecylazanium chloride
Synonyms: stearalkonium chloride
Molecular Weight (g/mol): 424.15
Molecular Formula: C27H50ClN
InChi Key: SFVFIFLLYFPGHH-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results