accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122-35-0

IUPAC Name: 2-phenoxypropanoyl chloride
Synonyms: propanoyl chloride, 2-phenoxy-
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: BDSSZTXPZHIYHM-UHFFFAOYNA-N
SMILES: CC(OC1=CC=CC=C1)C(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results