accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(Z)

CAS RN 122-40-7

IUPAC Name: (2Z)-2-(phenylmethylidene)heptanal
Synonyms: amyl cinnamic aldehyde amylcinnamaldehyde pentylcinnamaldehyde jasmine aldehyde α-amylcinnamaldehyde (natural) flomine amylcinnamic acid aldehyde jasminal jasminaldehyde α-amylcinnamicaldehyde
Molecular Weight (g/mol): 202.3
Molecular Formula: C14H18O
InChi Key: HMKKIXGYKWDQSV-KAMYIIQDSA-N
SMILES: CCCCC\C(C=O)=C\C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)