accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122-42-9

IUPAC Name: propan-2-yl N-phenylcarbamate
Synonyms: isopropyl phenyl carbamate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: VXPLXMJHHKHSOA-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)NC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results