accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(E)

CAS RN 122-57-6

IUPAC Name: (3E)-4-phenylbut-3-en-2-one
Synonyms: BENZ 2-phenylvinyl methyl ketone methyl styryl ketone benzylidene acetone methyl β-styryl ketone benzilideneacetone benzalacetone styryl methyl ketone methyl 2-phenylvinyl ketone 4-phenyl-3-buten-2-one
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: BWHOZHOGCMHOBV-BQYQJAHWSA-N
SMILES: CC(=O)\C=C\C1=CC=CC=C1
Boiling Point 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)