accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122-59-8

IUPAC Name: 2-phenoxyacetic acid
Synonyms: phenoxyacetic acid acide phenoxyacetique glycol acid phenyl ether POA phenoxyessigsaeure glycollic acid phenyl ether poa cpd o-phenylglycolic acid phenoxacetic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: LCPDWSOZIOUXRV-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (5)
  • (4)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (4)
  • (2)
  • (4)