accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOH

CAS RN 122-87-2

IUPAC Name: 2-[(4-hydroxyphenyl)amino]acetic acid
Synonyms: glycin N-phpg hydroxyphenyl glycine p-hydroxyphenyl glycine nphpg photoglycine D,L-(4-hydroxyphenyl)glycine
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: WRUZLCLJULHLEY-UHFFFAOYSA-N
SMILES: OC(=O)CNC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)