accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNOH

CAS RN 122-96-3

IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethan-1-ol
Synonyms: 1,4-piperazinediethanol
Molecular Weight (g/mol): 174.24
Molecular Formula: C8H18N2O2
InChi Key: VARKIGWTYBUWNT-UHFFFAOYSA-N
SMILES: OCCN1CCN(CCO)CC1
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)