accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122-98-5

IUPAC Name: 2-(phenylamino)ethan-1-ol
Synonyms: (+-)-phenylethanolamine
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: MWGATWIBSKHFMR-UHFFFAOYSA-N
SMILES: OCCNC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)