accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1220-00-4

IUPAC Name: 1,3-bis(4-chlorophenyl)thiourea
Synonyms: di-p-chlorophenylthiourea
Molecular Weight (g/mol): 297.2
Molecular Formula: C13H10Cl2N2S
InChi Key: SYJZYFOTCABQES-UHFFFAOYSA-N
SMILES: ClC1=CC=C(NC(=S)NC2=CC=C(Cl)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results