CAS RN 1220-78-6
IUPAC Name:
2-amino-4-[2-(3-amino-4-hydroxyphenyl)propan-2-yl]phenol
Synonyms:
2-amino-4-[2-(3-amino-4-hydroxyphenyl)propan-2-yl]phenol
Molecular Weight (g/mol):
258.32
Molecular Formula:
C15H18N2O2
InChi Key:
UHIDYCYNRPVZCK-UHFFFAOYSA-N
SMILES:
CC(C)(C1=CC=C(O)C(N)=C1)C1=CC=C(O)C(N)=C1
1
–
1
of
1
results
1
2,2-Bis(3-amino-4-hydroxylphenyl)propane 97.0+%, TCI America™
CAS: 1220-78-6 Molecular Formula: C15H18N2O2 Molecular Weight (g/mol): 258.32 MDL Number: MFCD00437413 InChI Key: UHIDYCYNRPVZCK-UHFFFAOYSA-N PubChem CID: 223319 IUPAC Name: 2-amino-4-[2-(3-amino-4-hydroxyphenyl)propan-2-yl]phenol SMILES: CC(C)(C1=CC=C(O)C(N)=C1)C1=CC=C(O)C(N)=C1