accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122049-54-1

IUPAC Name: 2-oxo-3-phenylpropanoate
Synonyms: β-phenylpyruvate calcium salt phenylpyruvic acid phenylpyroracemic acid phenylpyroracemate phenylpyruvate (substance) keto-phenylpyruvate sodium salt phenylpyruvic acid phenylpyruvic acid (substance)
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H7O3
InChi Key: BTNMPGBKDVTSJY-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(=O)CC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results