accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OOSnOONaNa

CAS RN 12209-98-2

IUPAC Name: disodium oxostannanebis(olate) trihydrate
Synonyms: disodium oxostannanebis(olate) trihydrate
Molecular Weight (g/mol): 266.73
Molecular Formula: H6Na2O6Sn
InChi Key: SFXJSNATBHJIDS-UHFFFAOYSA-N
SMILES: O.O.O.[Na+].[Na+].[O-][Sn]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)