CAS RN 122107-04-4
CAS RN 122107-04-4
IUPAC Name:
3,4-dihexylthiophene
Synonyms:
3,4-dihexylthiophene
Molecular Weight (g/mol):
252.46
Molecular Formula:
C16H28S
InChi Key:
YIRIIAIZQBBXHL-UHFFFAOYSA-N
SMILES:
CCCCCCC1=CSC=C1CCCCCC
1
–
1
of
1
results
1
3,4-Dihexylthiophene 97.0+%, TCI America™

CAS: 122107-04-4 Molecular Formula: C16H28S Molecular Weight (g/mol): 252.46 MDL Number: MFCD21363048 InChI Key: YIRIIAIZQBBXHL-UHFFFAOYSA-N PubChem CID: 15360364 IUPAC Name: 3,4-dihexylthiophene SMILES: CCCCCCC1=CSC=C1CCCCCC