accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1221237-87-1

N

CAS RN 1221237-87-1

IUPAC Name: 9-{[1,1'-biphenyl]-3-yl}-9H-carbazole
Synonyms: 9-{[1,1'-biphenyl]-3-yl}carbazole
Molecular Weight (g/mol): 319.41
Molecular Formula: C24H17N
InChi Key: LKXFMLDAUIXMGY-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC(=CC=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results