accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1221715-80-5

H3CBrFOH

CAS RN 1221715-80-5

IUPAC Name: 1-(3-bromo-2-fluorophenyl)ethan-1-ol
Synonyms: 1-(3-bromo-2-fluorophenyl)ethanol
Molecular Weight (g/mol): 219.05
Molecular Formula: C8H8BrFO
InChi Key: QFVPIDFXSBXICP-UHFFFAOYNA-N
SMILES: CC(O)C1=C(F)C(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results