accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOHOO

CAS RN 1221793-43-6

IUPAC Name: 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-4-oxopiperidine-2-carboxylic acid
Synonyms: 1-[(9H-fluoren-9-ylmethoxy)carbonyl]-4-oxopiperidine-2-carboxylic acid
Molecular Weight (g/mol): 365.39
Molecular Formula: C21H19NO5
InChi Key: YGAHCGORXONSSM-UHFFFAOYNA-N
SMILES: OC(=O)C1CC(=O)CCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results