accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122215-84-3

IUPAC Name: N-{[1,1'-biphenyl]-4-yl}-N-phenyl-[1,1'-biphenyl]-4-amine
Synonyms: N-{[1,1'-biphenyl]-4-yl}-N-phenyl-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 397.52
Molecular Formula: C30H23N
InChi Key: VXKVZTGWNITVPY-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results