accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122235-70-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 426.47
Molecular Formula: C23H26N2O6
InChi Key: MVWPBNQGEGBGRF-IBGZPJMESA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results