accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNNHN

CAS RN 1223020-29-8

IUPAC Name: 2,6-bis(1-benzyl-4,5-diphenylimidazolidin-2-yl)pyridine
Synonyms: 2,6-bis(1-benzyl-4,5-diphenylimidazolidin-2-yl)pyridine
Molecular Weight (g/mol): 703.93
Molecular Formula: C49H45N5
InChi Key: XLWFDEFJSJPDND-UHFFFAOYNA-N
SMILES: C(N1C(NC(C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC(=N1)C1NC(C(N1CC1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results