accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBrN

CAS RN 122306-01-8

IUPAC Name: (6-bromopyridin-3-yl)methanol
Synonyms: (6-bromopyridin-3-yl)methanol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: QPPDKOIDAYZUHN-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Br)N=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)