accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSiNSNSSiH3CH3CH3CCH3H3CCH3BrBrH3CH3CCH3CH3

CAS RN 1223559-98-5

IUPAC Name: 4-bromo-5-{4-bromo-2-[tris(propan-2-yl)silyl]-1,3-thiazol-5-yl}-2-[tris(propan-2-yl)silyl]-1,3-thiazole
Synonyms: 4-bromo-5-[4-bromo-2-(triisopropylsilyl)-1,3-thiazol-5-yl]-2-(triisopropylsilyl)-1,3-thiazole
Molecular Weight (g/mol): 638.71
Molecular Formula: C24H42Br2N2S2Si2
InChi Key: HNWAYAXELMAWTB-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)C1=NC(Br)=C(S1)C1=C(Br)N=C(S1)[Si](C(C)C)(C(C)C)C(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results