CAS RN 122433-41-4
IUPAC Name:
methyl 5-bromo-2-methoxypyridine-3-carboxylate
Synonyms:
methyl 5-bromo-2-methoxypyridine-3-carboxylate
Molecular Weight (g/mol):
246.06
Molecular Formula:
C8H8BrNO3
InChi Key:
OLQOVEOEHQDKSC-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Br)=CN=C1OC
1
–
1
of
1
results
1
Sigma Aldrich Methyl 5-bromo-2-methoxynicotinate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.