CAS RN 1224430-39-0
IUPAC Name:
4-hexylthiophene-2-carbonitrile
Synonyms:
4-hexylthiophene-2-carbonitrile
Molecular Weight (g/mol):
193.31
Molecular Formula:
C11H15NS
InChi Key:
LFLOKABSZNAYLC-UHFFFAOYSA-N
SMILES:
CCCCCCC1=CSC(=C1)C#N
1
–
1
of
1
results
1
4-Hexylthiophene-2-carbonitrile 95.0+%, TCI America™
CAS: 1224430-39-0 Molecular Formula: C11H15NS Molecular Weight (g/mol): 193.308 MDL Number: MFCD18413940 InChI Key: LFLOKABSZNAYLC-UHFFFAOYSA-N Synonym: 2-Cyano-4-hexylthiophene PubChem CID: 58420906 IUPAC Name: 4-hexylthiophene-2-carbonitrile SMILES: CCCCCCC1=CSC(=C1)C#N