CAS RN 12245-39-5
CAS RN 12245-39-5
IUPAC Name:
λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2Z)-4-oxopent-2-en-2-olate
Synonyms:
λ¹-rhodium(1+) 1,5-cyclooctadiene, (Z,Z)- (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol):
310.2
Molecular Formula:
C13H19O2Rh
InChi Key:
TVGPAHONTUMCNP-DWVXZKBMSA-M
SMILES:
[Rh+].C\C([O-])=C\C(C)=O.C1C\C=C/CC\C=C/1
1
–
1
of
1
results
1
Sigma Aldrich 2-Mesityl-5-Methylimidazo[1,5-A]Pyridinium Chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.