accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSnH3CCH3NNSOOCH3

CAS RN 122476-85-1

IUPAC Name: 2-methanesulfonyl-5-(tributylstannyl)pyrimidine
Synonyms: 2-methanesulfonyl-5-(tributylstannyl)pyrimidine
Molecular Weight (g/mol): 447.23
Molecular Formula: C17H32N2O2SSn
InChi Key: RRHTVQOGUAOHDR-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CN=C(N=C1)S(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results