accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHHN

CAS RN 122536-76-9

IUPAC Name: tert-butyl N-(pyrrolidin-3-yl)carbamate
Synonyms: tert-butyl N-(pyrrolidin-3-yl)carbamate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: DQQJBEAXSOOCPG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCNC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)