CAS RN 122547-49-3
CAS RN 122547-49-3
IUPAC Name:
sodium (5R,6S)-6-[(1R)-1-hydroxyethyl]-7-oxo-3-[(2R)-oxolan-2-yl]-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
Synonyms:
sodium (5R,6S)-6-[(1R)-1-hydroxyethyl]-7-oxo-3-[(2R)-oxolan-2-yl]-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
Molecular Weight (g/mol):
307.3
Molecular Formula:
C12H14NNaO5S
InChi Key:
ICSAXRANXQSPQP-VUKDEKJYSA-M
SMILES:
[Na+].[H][C@]12SC(=C(N1C(=O)[C@]2([H])[C@@H](C)O)C([O-])=O)[C@@]1([H])CCCO1
1
–
1
of
1
results
1
Faropenem sodium hydrate, Thermo Scientific Chemicals
CAS: 122547-49-3 Molecular Formula: C12H14NNaO5S·xH2O Molecular Weight (g/mol): 307.3