accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONHNNNOOH3CCH3H3COOCH3H3CCH3

CAS RN 122555-91-3

IUPAC Name: tert-butyl 2-{4,7-bis[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
Synonyms: tert-butyl 2-{4,7-bis[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
Molecular Weight (g/mol): 514.71
Molecular Formula: C26H50N4O6
InChi Key: NMHVTLJFPDOJOD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CN1CCNCCN(CC(=O)OC(C)(C)C)CCN(CC(=O)OC(C)(C)C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results