CAS RN 122564-82-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(±)-Anatoxin A fumarate, Tocris Bioscience™
CAS: 122564-82-3 Molecular Formula: C10H15NO·C4H4O4 Synonym: (±)-2-Acetyl-9-aza bicyclo[4.2.1]non-2-ene fumarate