accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12257-42-0

IUPAC Name: bis(λ¹-rhodium(1+)) bis(bicyclo[2.2.1]hepta-2,5-diene) dichloride
Synonyms: bis(λ¹-rhodium(1+)) bis(norbornadiene) dichloride
Molecular Weight (g/mol): 460.99
Molecular Formula: C14H16Cl2Rh2
InChi Key: RXDWVIOULPVOEO-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Rh+].[Rh+].C1C2C=CC1C=C2.C1C2C=CC1C=C2
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)