CAS RN 122590-77-6
IUPAC Name:
4-azido-2,3,5,6-tetrafluorobenzoic acid
Synonyms:
4-azido-2,3,5,6-tetrafluorobenzoic acid
Molecular Weight (g/mol):
235.1
Molecular Formula:
C7HF4N3O2
InChi Key:
IOJFHZXQSLNAQJ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C(F)=C(N=[N+]=[N-])C(F)=C1F
1
–
1
of
1
results
1
4-Azido-2,3,5,6-tetrafluorobenzoic Acid 98.0+%, TCI America™
CAS: 122590-77-6 Molecular Formula: C7HF4N3O2 Molecular Weight (g/mol): 235.10 MDL Number: MFCD00467436 InChI Key: IOJFHZXQSLNAQJ-UHFFFAOYSA-N PubChem CID: 2762578 IUPAC Name: 4-azido-2,3,5,6-tetrafluorobenzoic acid SMILES: OC(=O)C1=C(F)C(F)=C(N=[N+]=[N-])C(F)=C1F