accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3SNCH3H3C

CAS RN 1226-46-6

IUPAC Name: 4-[4-(dimethylamino)benzenecarbothioyl]-N,N-dimethylaniline
Synonyms: 4-[4-(dimethylamino)benzenecarbothioyl]-N,N-dimethylaniline
Molecular Weight (g/mol): 284.42
Molecular Formula: C17H20N2S
InChi Key: KFUJUTFTRXYQMG-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(=S)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results