accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12263-33-1

IUPAC Name: trimanganese(2+) diphosphanetriide
Synonyms: trimanganese(2+) diphosphanetriide
Molecular Weight (g/mol): 226.76
Molecular Formula: Mn3P2
InChi Key: ZZDYLDYDQHMVJZ-UHFFFAOYSA-N
SMILES: [P-3].[P-3].[Mn++].[Mn++].[Mn++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results