accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12266-38-5

IUPAC Name: stibanylidyneplumbyl
Synonyms: stibanylidyneplumbyl
Molecular Weight (g/mol): 329
Molecular Formula: PbSb
InChi Key: QQHJESKHUUVSIC-UHFFFAOYSA-N
SMILES: [Sb]#[Pb]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results