accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPtH3CHCCHCHHC

CAS RN 12266-92-1

IUPAC Name: 1,1-dimethanuidyl-1-platinatetracyclo[4.3.0.0¹,⁵.0²,⁹]nonane-2,5,6,9-tetrauide
Synonyms: 1,1-dimethanuidyl-1-platinatetracyclo[4.3.0.0¹,⁵.0²,⁹]nonane-2,5,6,9-tetrauide
Molecular Weight (g/mol): 333.34
Molecular Formula: C10H18Pt
InChi Key: YITHLUKCACVSOM-UHFFFAOYNA-N
SMILES: [CH3-][Pt]123([CH3-])[CH-]4CC[CH-]1[CH-]2CC[CH-]34
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results