accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122666-87-9

IUPAC Name: disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate dihydrate
Synonyms: tyrosine disodium dihydrate
Molecular Weight (g/mol): 261.19
Molecular Formula: C9H13NNa2O5
InChi Key: WFTCFVUQORELJZ-CMIWSEPMNA-L
SMILES: O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)