accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHNOH3COOCH3OHNH3CCH3CH3ICH3

CAS RN 1226896-38-3

IUPAC Name: 2-(2-iodo-3-{1-[(2,4,6-trimethylphenyl)carbamoyl]ethoxy}phenoxy)-N-(2,4,6-trimethylphenyl)propanamide
Synonyms: 2-(2-iodo-3-{1-[(2,4,6-trimethylphenyl)carbamoyl]ethoxy}phenoxy)-N-(2,4,6-trimethylphenyl)propanamide
Molecular Weight (g/mol): 614.52
Molecular Formula: C30H35IN2O4
InChi Key: ZVOKSLMZXDIXPR-UHFFFAOYNA-N
SMILES: CC(OC1=CC=CC(OC(C)C(=O)NC2=C(C)C=C(C)C=C2C)=C1I)C(=O)NC1=C(C)C=C(C)C=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results