accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1227-44-7

IUPAC Name: 1,3-bis(4-methoxyphenyl)urea
Synonyms: 1,3-bis(4-methoxyphenyl)urea
Molecular Weight (g/mol): 272.3
Molecular Formula: C15H16N2O3
InChi Key: CPSIMDUNVYMPAH-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=O)NC2=CC=C(OC)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results