accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1227-45-8

IUPAC Name: 1,3-bis(4-methoxyphenyl)thiourea
Synonyms: 1,3-bis(4-methoxyphenyl)thiourea
Molecular Weight (g/mol): 288.37
Molecular Formula: C15H16N2O2S
InChi Key: RGRJEERVXALLTH-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=S)NC2=CC=C(OC)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results