accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1227301-51-0

IUPAC Name: N-[1-({7-[(4-aminobutyl)amino]heptyl}carbamoyl)-2-(4-hydroxyphenyl)ethyl]butanamide dihydrochloride
Synonyms: N-[1-({7-[(4-aminobutyl)amino]heptyl}carbamoyl)-2-(4-hydroxyphenyl)ethyl]butanamide dihydrochloride
Molecular Weight (g/mol): 507.54
Molecular Formula: C24H44Cl2N4O3
InChi Key: HWTJQQMIKVJWLH-UHFFFAOYNA-N
SMILES: Cl.Cl.CCCC(=O)NC(CC1=CC=C(O)C=C1)C(=O)NCCCCCCCNCCCCN
Product Line 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results