accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHNNHCH3

CAS RN 122734-32-1

IUPAC Name: tert-butyl N-[2-(methylamino)ethyl]carbamate
Synonyms: tert-butyl N-[2-(methylamino)ethyl]carbamate
Molecular Weight (g/mol): 174.24
Molecular Formula: C8H18N2O2
InChi Key: GKWGBMHXVRSFRT-UHFFFAOYSA-N
SMILES: CNCCNC(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)