accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1227579-04-5

IUPAC Name: 2,3-difluoro-6-methylpyridine
Synonyms: 2,3-difluoro-6-methylpyridine
Molecular Weight (g/mol): 129.11
Molecular Formula: C6H5F2N
InChi Key: HGBWAUANMSZOIJ-UHFFFAOYSA-N
SMILES: CC1=NC(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results