CAS RN 1227628-83-2
IUPAC Name:
2-fluoro-4-(trifluoromethoxy)benzaldehyde
Synonyms:
2-fluoro-4-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol):
208.11
Molecular Formula:
C8H4F4O2
InChi Key:
DEACWPDDLIOZJV-UHFFFAOYSA-N
SMILES:
FC1=C(C=O)C=CC(OC(F)(F)F)=C1
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%
2-Fluoro-4-(trifluoromethoxy)benzaldehyde, 95%
CAS: 1227628-83-2 Molecular Formula: C8H4F4O2 Molecular Weight (g/mol): 208.112 MDL Number: MFCD16652440 InChI Key: DEACWPDDLIOZJV-UHFFFAOYSA-N Synonym: 2-fluoro-4-trifluoromethoxy benzaldehyde PubChem CID: 53465593 IUPAC Name: 2-fluoro-4-(trifluoromethoxy)benzaldehyde SMILES: C1=CC(=C(C=C1OC(F)(F)F)F)C=O