accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12277-88-2

IUPAC Name: (1Z,5Z)-cycloocta-1,5-diene; diphenylplatinumbis(ylium)
Synonyms: 1,5-cyclooctadiene, (Z,Z)-; diphenylplatinumbis(ylium)
Molecular Weight (g/mol): 457.48
Molecular Formula: C20H22Pt
InChi Key: MQCNDJYHICNUPD-PHFPKPIQSA-N
SMILES: C1C\C=C/CC\C=C/1.[Pt++](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results