accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOI

CAS RN 122775-34-2

IUPAC Name: 3-iodo-4H-chromen-4-one
Synonyms: 3-iodochromen-4-one
Molecular Weight (g/mol): 272.04
Molecular Formula: C9H5IO2
InChi Key: SZDBDDKQJHHRGH-UHFFFAOYSA-N
SMILES: IC1=COC2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results